Corning® CellSTACK® 10 Chamber with Vent Caps, Corning | Avantor
Corning® CellSTACK® 10 Chamber with Vent Caps, Corning
Polystyrene CellSTACK® - 10 Chamber with Vent Caps, Polystyrene, 6360 cm2, Sterile
Catalog # 89094-692
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  7073
Corning® CellSTACK® 10 Chamber with Vent Caps, Corning
Catalog # 89094-692
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Corning® CellSTACK® 10 Chamber with Vent Caps, Corning
Corning® CellSTACK® 10 Chamber with Vent Caps, Corning
Polystyrene CellSTACK® - 10 Chamber with Vent Caps, Polystyrene, 6360 cm2, Sterile
Price
$3,119.50
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 6
Product Details & Documents
Has 6360 cm² cell growth area, two 26 mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system, Standard 33 mm threaded caps have 0.2 µm pore nonwettable membranes sealed directly to the caps to allow gas exchange while minimizing the risk of contamination, Optional 33 mm threaded caps are available with integrally sealed chemically resistant.
Heat sealable flexible tubing to allow direct sterile transfer of media and cells, USP Class VI polystyrene provides excellent optical clarity and mechanical strength, sterilized by gamma radiation and certified nonpyrogenic, 100% leak tested prior to shipping.
Rectangular flask shape.
Spécifications
  • Désignation:
    Polystyrene CellSTACK® - 10 Chamber with Vent Caps
  • Matière:
    Polystyrene
  • Growth Area:
    6360 cm2
  • Sterility:
    Sterile
  • Cat. No.:
    89094-692
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Media volume:
    1250 - 1900 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Surface:
    Tissue Culture Treated