Corning® 25-Layer CellCube® Module, Corning
À propos de cet article
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
1 Options Available Below
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Specifications for Product Family
Spécifications
- Catalog No:
- 490014-826
- 89090-254
- Environmentally Preferable:
- Low Manufacturing Impact
- Media volume:
- 1500 ml
- 1500 ml
- Surface:
- Corning® CellBIND®
- Tissue Culture Treated
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
Spécifications
Product Family Options
Informations sur le produit
- DésignationMatièreGrowth AreaSterilityAvailabilityPrice
- 25-Layer CellCube® Module with 21,250 cm² Growth SurfacePolycarbonate/Polystyrene21,250 cm²SterileDiscontinued-Supplier Number: 3304Spécifications:
More
Specifications
Cat. No.490014-826Environmentally PreferableMedia volume1500 mlSurfaceCorning® CellBIND®Environmental certification - 25-Layer CellCube® Module with 21,250 cm² Growth SurfacePolycarbonate/Polystyrene21,250 cm²SterileLog in pour la disponibilitéCase of 1$1,260.00Supplier Number: 3201Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89090-254Environmentally PreferableLow Manufacturing ImpactMedia volume1500 mlSurfaceTissue Culture TreatedEnvironmental certificationRECs, ISO 14001 - Kennebunk