Extraction Thimbles, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These extraction thimbles, made from cellulose fiber, provide good retention
They are seamless, high quality, single thickness. Readily permeable to the flow of ether and other organic solvents.
Frequently Bought Together