ContactUsLinkComponent
Visual Scientifics Pendulum
  470135-570
 :  Wards
undefined
Visual Scientifics Pendulum
  470135-570
 :  Wards
 :  PTPEN
  • Description:
    Visual scientifics pendulum
  • Cat. no.:
    470135-570
  • Supplier No.:
    PLPEN
Discover the period of a simple pendulum, then use video analysis to investigate energy conservation and conversion.
  • 20-page lab manual included, with activities and extensions
  • Covers 10 NGSS and 8 common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
Students can apply technology to study the Law of Conservation of Energy and energy conversions using this engaging pendulum system. The included lab manual provides approximately three hours of laboratory activities, and meets multiple Next Generation Science Standards.
 : Visual Scientifics Base (470148-822) required, but sold separately.
 : This product inlcudes a pendulum backboard, mounting rod, pendulum bob and stopper, and string. Video camera, graphing software, and photogate system are recommended.