Visual Scientifics Loop Track | Avantor
Visual Scientifics Loop Track
Visual scientifics loop track
Catalog # 470135-568
Supplier:   Wards
Fournisseur #:  PTLOP
Visual Scientifics Loop Track
Catalog # 470135-568
Supplier:  Wards
Visual Scientifics Loop Track
Visual Scientifics Loop Track
Visual scientifics loop track
Price
$46.35
Log in pour la disponibilité
Each
Product Details & Documents
Students apply concepts of conservation of energy to a loop setup, then consider the forces involved in roller coasters.
  • 30-page lab manual included, with activities and extensions
  • Covers 15 NGSS and 17 common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
Students investigate the conversion between kinetic and potential energy and explore energy conservation. Using video analysis, students can investigate normal force on a roller coaster rider. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and common core concepts.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately. This kit includes a metal ramp with loop, and two metal balls. Recommended accessories: video camera and graphical analysis software, scale, and ruler or measuring tape.
Spécifications
  • Désignation:
    Visual scientifics loop track
Accessories
Informations sur le produit
  • Catalog #
    Size
    Availability
    Price
  • Visual scientifics base
    Visual scientifics base
    470148-822
    25×16×2.5"
    Log in pour la disponibilité
    Each - $214.85