Visual Scientifics Loop Track
Product Details & Documents
Students apply concepts of conservation of energy to a loop setup, then consider the forces involved in roller coasters.
- 30-page lab manual included, with activities and extensions
- Covers 15 NGSS and 17 common core standards
- Platform neutral - works with your preferred probeware/video analysis tools
Students investigate the conversion between kinetic and potential energy and explore energy conservation. Using video analysis, students can investigate normal force on a roller coaster rider. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and common core concepts.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately. This kit includes a metal ramp with loop, and two metal balls. Recommended accessories: video camera and graphical analysis software, scale, and ruler or measuring tape.
Spécifications
- Désignation:Visual scientifics loop track
Accessories
Informations sur le produit
- Catalog #SizeAvailabilityPrice
- Visual scientifics base
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.