ContactUsLinkComponent
Visual Scientifics Collisions in Two Dimensions
Catalog # 470135-564
Supplier:  Wards
undefined
Visual Scientifics Collisions in Two Dimensions
Catalog # 470135-564
Supplier:  Wards
Supplier Number:  PT2SCO
Specifications
  • Description:
    Visual scientifics collisions in two dimensions
  • Cat. no.:
    470135-564
  • Supplier No.:
    PT2SCO
Specifications
About this item
Teach students about two-dimensional collisons with this engaging activity, then use video analysis to analyze conservation of energy, and conversions between kinetic and potential energy.
  • 28 page lab manual included, with activities and extensions
  • Covers 12 NGSS and 13 common core standards
  • Platform Neutral - works with your preferred probeware/video analysis tools
Students can apply technology to study the Law of Conservation of Energy and energy conversions, as well as momentum and elastic collisions. The included lab manual provides approximately three hours of laboratory activities, and meets multiple Next Generation Science Standards.
Ordering information: Visual Scientifics Base (470148-822) and Backboard (470135-558) required, but sold separately.
Delivery: This kit includes a metal ramp, 2 metal balls and 1 glass ball. The activity requires the Visual Scientifics Base and Backboard, which are available separately. Recommended accessories: video camera, graphing software, carbon paper, scale, protractor, and ruler/measuring tape.