Puritan® Applicator, Cotton Tip, Polystyrene Shaft, Puritan Medical Products
Catalog # 89218-846
Supplier: Puritan Medical Products
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Description:Polystyrene Shaft
- Cat. no.:89218-846
- Tip Size:17.4 mm (0.687")
- Handle Length:126.23 mm (4.970")
- Tip Diameter:5.15 mm (0.203")
- Handle Diameter:2.54 mm (0.1")
- Supplier No.:25-805
Specifications
About this item
These applicators are DNA controlled with a special patented tube design for product breathability, sterile
These have with a standard tip.
They are manufactured in a DNA controlled an monitored clean room environment.