Puritan® Histobrush® Cytology Collection Devices, Puritan Medical Products
Puritan® cytology brushes feature tapered Histobrush® tips made out of soft nylon bristles, attached to a long polystyrene handle
They are used to collect cervical specimens during pap smear tests, as well as buccall cells for DNA lineage.
: CE marked for medical use. Made in the USA.
: Sterile brushes are individually wrapped.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together