Puritan® Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
These knitted polyester swabs are non-abrasive and lint-free
Ideal for instrumentation and medical device manufacturing. Made in the USA.
In addition, the polyester can easily stand up to IPA, acetone, and other solvents commonly used in clean rooms and other critical environments.
Certifications: CE marked.
Ordering information: Swabs are packaged 100 per pack, 10 packs per case.
Frequently Bought Together