PurSwab® Pointed Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Pointed foam tip is heat staked to a green polypropylene handle and fits in small tight areas
Made in the USA.
Used for precision cleaning. Can be used with IPA and DI water. Lint- and residue-free.
: CE marked.
: Packaged as 10 bags if 100 swabs.
Frequently Bought Together