PurSwab® Small Paddle Shaped Foam Tipped Applicator, Black Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Certificates
About this item
These swabs have thermal edge seals and thermal bonding to the handle help ensure the sponge-like foam will clean up unwanted particles without releasing any into your critical environment.
Small flat paddle that is lint and residue free.
Made in the USA.
Certifications: CE marked.
Ordering information: Packaged as 2 bags of 500 swabs per bag.
Frequently Bought Together