VWR® Upright Freezer Racks for 96 Deep Well Microtiter Plates, Stainless Steel
Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.
- Stainless steel construction
- Sturdy handle design
- Easy to reorder
- Standard identification labels
Racks feature sturdy handles and come standard with identification labels for inventory purposes. Racks accommodate 96 deep well microtiter plates and contain locking rods. Reorder by simply scanning the QR code with your smartphone or tablet.
Frequently Bought Together