Socorex Acura® Single-Channel Pipettors, Variable Volume, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Acura® manual 826 micropipettes designed with the research scientist in mind.
- Shorter, smoother, and lightweight, the eight models offer key features having excellent ergonomics
- XS (eXtra Sharp) pipette features a shorter shaft making it easier than ever to dispense and aspirate into the smallest of microtubes
The Acura is very lightweight (<88 grams) helps eliminate fatigue during long hours of pipetting. The ultra-soft plunger greatly reduces thumb and hand fatigue for added comfort and safety.
The Swift-Set Calibration provides easy user calibration system keeping instrument tuned for accurate pipetting. The Acura XS micropipettes are Autoclavable fully assembled and provides an easy way to eliminate cross contamination.
Ordering information: Each pipette also includes tip samples, QC certificate, and operating instructions. The Acura® 826 XS Pipette Starter Kit includes one 826 XS (0.5 to 10 µl), one 826 XS (10 to 100 µl), one 826 XS (20 to 200 µl), one 826 XS (100 to 1000 µl), one Twister Blue Pipette Stand, one rack with 10 µl tips, two racks with 200 µl tips, and one rack with 1000 µl tips.