Contrex® CA Acidic Liquid Detergent
3.8 L (1 gal.), Poly bottle
Supplier: Decon Labs
Fournisseur #: 5701
Contrex® CA Acidic Liquid Detergent
Catalog # CA89234-834
Supplier: Decon Labs
Jump to:
Product Details & Documents
Contrex® CA concentrated acidic detergent is formulated for use in descaling, mineral removal, oxide removal, or in applications where an acidic detergent is indicated.
Completely free-rinsing and biodegradable
Phosphate-free and disposable straight to drain
Contrex® CA contains a safe, organic acid and is phosphate-free, so it will not interfere with sensitive equipment or testing procedures.
Spécifications
- Taille:3.8 L (1 gal.)
- Packaging:Poly bottle
- Cat. No.:CA89234-834
- Environmentally Preferable:Sustainable Packaging|Waste Reducing