Contrex® EZ Powder Detergent
Fournisseur: Decon Labs
Certificats
À propos de cet article
Contrex® EZ is a concentrated, powdered enzymatic detergent.
Prevents corrosion
- Free-rinsing and disposable straight to drain
Contrex® EZ is formulated for use as an ezymatic detergent for use in manual or soak cleaning of labware and equipment contaminated with protein and fats.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together