Contrex® AP Powder Detergent | Avantor
Contrex® AP Powder Detergent
Contrex® AP Powder Detergent
Supplier:  Decon Labs
Certificates
Contrex® AP Powder Detergent
Contrex® AP Powder Detergent
2 Options Available Below
Product Details & Documents
Contrex® AP is a powdered labware detergent.
  • Biodegradable and readily disposable
  • User friendly bottle allows for easy dispensing and eliminates caking
Contrex® AP removes virtually any contaminant including grease, oils, blood, tissue, particulates, deposits, chemicals, and solvents. It works by soaking, scrubbing, or in ultrasonic cleaning baths. The totally rinsable formulation eliminates detergent build-up that interferes with analysis, tissue culture, and other critical procedures. Its activity can be accelerated by heating, agitation, and ultrasonics, or by increasing concentrations.
Documents
Product Family Options
Informations sur le produit
  • Taille
    Packaging
    Availability
    Price
  • 1.8 Kg (4 lbs.)
    Poly bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $70.40
    $70.40
    Log in pour la disponibilité
    Case of 9 (1/ Itm/Bx)
    Case of 9 (1/ Itm/Bx) - $633.60
    $633.60
    Each(1/Itm/Bx)
       $70.40
    Case of 9 (1/ Itm/Bx)
       $633.60
    Supplier Number: 5204
  • 11.3 Kg (25 lbs.)
    Poly bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (25lb)
    Each (25lb) - $373.00
    $373.00
    Each (25lb)
       $373.00
    Supplier Number: 5225