Conflikt® Ready-to-use Disinfectant
Supplier: Decon Labs
Certificates
About this item
Conflikt ® Ready-to-use disinfectant spray cleans and disinfects hard surfaces that may harbor viruses, bacteria and fungi.
Cleans and disinfects hard surfaces that may harbor viruses, bacteria, and fungi
- Effective against a wide range of organisms
- Conflikt® spray is approved by USDA
Conflikt® spray is especially formulated to disinfect hard, non-porous surfaces such as glass, plastics, stainless steel, glazed ceramics, laminates, metal surfaces, glazed porcelain, etc.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together