Contrad® 70 Liquid Detergent | Avantor
Contrad® 70 Liquid Detergent
   Options durables disponibles
Supplier:   Decon Labs
Contrad® 70 Liquid Detergent
Supplier:  Decon Labs
   Options durables disponibles
Certificates
Contrad® 70 Liquid Detergent
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Contrad® 70 is an emulsion of anionic and non-ionic surfactants in an alkaline aqueous base.
  • Phosphate-free, chlorine-free, and biodegradable
  • Free-rinsing and disposable straight to the drain
  • Cleans glass, plastic, ceramic, and ferrous metals
Contrad® 70 works by soaking, scrubbing, or in ultrasonic cleaning baths. It removes virtually any contaminant from every surface, even silicones, dried proteins, carbon, most resins, waxes, and oils. The formulation is totally rinsable, eliminating detergent build-up that interferes with analysis, tissue culture, and other critical procedures. It is also extremely effective when used as an ambient soak solution, with no need to scrub. Its activity can be accelerated by heating, agitation, and ultrasonics, or by increasing concentrations. Used solutions may be disposed straight to drain providing they have not been used to remove contaminants that are in themselves pollutants.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 42000-000
  • CA42000-000
  • CA89233-154
  • CA89233-152
  • CA89233-156
  • Environmentally Preferable:
  • Sustainable Packaging|Waste Reducing
  • Sustainable Packaging|Waste Reducing
  • Sustainable Packaging|Waste Reducing
  • Sustainable Packaging|Waste Reducing
  • Sustainable Packaging|Waste Reducing
Spécifications
Product Family Options
Informations sur le produit
  • Taille
    Packaging
    Availability
    Price
  • 5 L ( 1.32 gal.)
    Poly bottle
    Discontinued
    Supplier Number: 1003
    Spécifications:
      Sustainable Packaging|Waste Reducing
    More
    Specifications
    Cat. No.
    42000-000
    Environmentally Preferable
    Sustainable Packaging|Waste Reducing
      Sustainable Packaging|Waste Reducing
  • 5 L ( 1.32 gal.)
    Poly bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $236.00
    $236.00
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $944.00
    $944.00
    Each
       $236.00
    Case of 4
       $944.00
    Supplier Number: 1003
    Spécifications:
      Sustainable Packaging|Waste Reducing
    More
    Specifications
    Cat. No.
    CA42000-000
    Environmentally Preferable
    Sustainable Packaging|Waste Reducing
      Sustainable Packaging|Waste Reducing
  • 20 L (5.2 gal.)
    Poly drum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20l)
    Each (20l) - $935.00
    $935.00
    Each (20l)
       $935.00
    Supplier Number: 1004
    Spécifications:
      Sustainable Packaging|Waste Reducing
    More
    Specifications
    Cat. No.
    CA89233-154
    Environmentally Preferable
    Sustainable Packaging|Waste Reducing
      Sustainable Packaging|Waste Reducing
  • 1 L (34 oz.)
    Poly bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1l)
    Each(1l) - $74.40
    $74.40
    Log in pour la disponibilité
    Case of 12 (1 l)
    Case of 12 (1 l) - $892.80
    $892.80
    Each(1l)
       $74.40
    Case of 12 (1 l)
       $892.80
    Supplier Number: 1002
    Spécifications:
      Sustainable Packaging|Waste Reducing
    More
    Specifications
    Cat. No.
    CA89233-152
    Environmentally Preferable
    Sustainable Packaging|Waste Reducing
      Sustainable Packaging|Waste Reducing
  • 210 L (55 gal.)
    Poly drum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (210l)
    Each (210l) - $6,190.00
    $6,190.00
    Each (210l)
       $6,190.00
    La quantité minimale de commande est de 1
    Supplier Number: 1005
    Spécifications:
      Sustainable Packaging|Waste Reducing
    More
    Specifications
    Cat. No.
    CA89233-156
    Environmentally Preferable
    Sustainable Packaging|Waste Reducing
      Sustainable Packaging|Waste Reducing