BART Test for Heterotropic Aerobic Bacteria, Hach
Catalogue # CA97026-678
Fournisseur: Hach
Numéro CAS:
Spécifications
- Cat. no.:CA97026-678
- Supplier No.:2490409
Spécifications
À propos de cet article
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Heterotrophic Aerobic Bacteria (HAB) are one of the most important agents in biofouling, causing corrosion, clogging, fouling of the water, and increased hygiene risks.
Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria.
Easy to use, requiring no elaborate or costly equipment and no specialized training. Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment.