pH Paper, Hach®
100, 0 - 14
Supplier: Hach
Fournisseur #: 2601300
Jump to:
Product Details & Documents
Provides a simple, fast, economical method of pH measurement
Obtain accurate results in weakly buffered, turbid and lightly colored samples.
Spécifications
- pH Range:0 - 14
- No. of Tests:100