pH Test Strips, Hach®
50, pH 4 - 9
Catalog # CA11014-386
Supplier: Hach
Supplier #: 2745650
Jump to:
Product Details & Documents
Most natural waters range from pH 4 to pH 9, but commonly are above pH 7 because of carbonates and bicarbonates (alkalinity). The pH of water used in industry, boiler feed water, and swimming pools usually is kept within a narrow range.
Easy to use, disposable, and inexpensive.
A great way to obtain quick, quantitative answers in the field or in the lab.
Specifications
- pH Range:pH 4 - 9
- No. of Tests:50