Flask, Reaction, Flat Flange, O-RIng Groove, Round Bottom, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
À propos de cet article
Cylindrical, heavy-wall reaction flask, round bottom
Top has a flat flange with an o-ring groove for use with CAPFE® (PTFE encapsulated silicone rubber) o-ring instead of a gasket. Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Fabricated from borosilicate glass.
Ordering information: Flask is supplied with one CAPFE o-ring, 89061-176 for 1000mL size, 89061-178 for 2000, 3000 and 4000mL sizes.
Frequently Bought Together