Flask, Dewar, Low Form, Ace Glass Incorporated
This cylindrical, low form flask is silvered and evacuated
Ideal for sub-ambient work. The low form design allows use of multi-neck flasks with magnetic stirring.
- Catalog No:
- 89057-566
- 89057-564
- 89057-570
- 89057-574
- Capacity:
- 0.35 L
- 0.15 L
- 0.85 L
- 2.5 L
- Int.Ø:
- 80 mm
- 80 mm
- 130 mm
- 170 mm
- Int. height:
- 75 mm
- 35 mm
- 75 mm
- 135 mm
Frequently Bought Together