Guardian™ Airflow Monitor, Labconco®
Catalog # 89233-240
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Guardian™ Airflow Monitor, 230V, 50/60Hz
- Cat. no.:89233-240
- REDISHIP:No
- Supplier No.:9413301
Specifications
About this item
Guardian Airflow Monitors are designed to continuously monitor airflow through Labconco fume hoods
Provides visual and audible alarms for abnormal airflow conditions.