Economical alternative to ultransonic disruptors.
- Rapid hands-free disruption of up to 12 microtubes (1.5 mL or 2.0 mL) at a time
- Use with Disruptor Beads, made from spherical, lead free, soda lime glass
- Suitable for use in cold rooms or incubators
- Supplied with Microtube holder for 12 microtubes (tubes not included), and Pop-Off Cup
- Adjustable digital speed alarm
- 2-year warranty included
Dramatically increase cell disruption efficiency
This Disruptor is available in analog or digital models, allows for Hands-Free disruption of up to twelve 1.5 or 2.0ml microtubes with timer from 0-15 minutes or continuous for analog models, and from 0-99 minutes or continuous for Digital models. Provides multi-directional action, which simultaneously agitates and vortexes at high speed, dramatically increasing cell disruption or sample resuspension efficiency. Suitable for use in cold rooms or incubators. Ideal for difficult glass bead procedures - cell disruption/homogenization of yeast, bacteria, plant and animal tissue or anything that requires extremely violent high speed agitation. Digital model incorporates an adjustable speed alarm. Remove microtube holder and attach the supplied Pop-off Cup for high speed vortexing of single test tubes.
The patented Disruptor Genie provides multi-directional action, which simultaneously agitates and vortexes at high speed, dramatically increasing cell disruption or sample resuspension effeciency.
- Catalog No:
- 89202-318
- 490010-034
- 490010-032
- 89202-324
- 89202-320
- 89202-316
- 89202-314
- Speed range:
- 3000 rpm (Analog), 1000-3000 rpm (Digital)
- 3000 rpm (Analog), 1000-3000 rpm (Digital)
- 3000 rpm (Analog), 1000-3000 rpm (Digital)
- Time range:
- 0-15 minutes (Analog), 0-99 minutes (Digital)
- 0-15 minutes (Analog), 0-99 minutes (Digital)
- 0-15 minutes (Analog), 0-99 minutes (Digital)
- Dimensions:
- 165D x 122W x 190H mm (61/2 x 44/5 x 71/2")
- 6.5" x 4.8" x 7.5" (165 x 122 x 190 mm)
- 6.5" x 4.8" x 7.5" (165 x 122 x 190 mm)
- 6.5" x 4.8" x 7.5" (165 x 122 x 190 mm)
- 165D x 122W x 190H mm (61/2 x 44/5 x 71/2")
- 165D x 122W x 190H mm (61/2 x 44/5 x 71/2")
- 165D x 122W x 190H mm (61/2 x 44/5 x 71/2")
- Weight:
- 4.3 Kg (91/2 lbs.)
- 9.5 lbs. (4.3 kg)
- 9.5 lbs. (4.3 kg)
- 9.5 lbs. (4.3 kg)
- 4.3 Kg (91/2 lbs.)
- 4.3 Kg (91/2 lbs.)
- 4.3 Kg (91/2 lbs.)
- Capacity:
- 12 microtubes, 1.5 or 2.0ml, snap-top or screw-on
- 12 microtubes, 1.5 or 2.0ml, snap-top or screw-on
- 12 microtubes, 1.5 or 2.0ml, snap-top or screw-on
- 12 microtubes, 1.5 or 2.0ml, snap-top or screw-on