ContactUsLinkComponent
Gel/PCR DNA Fragment Extraction Kits (Large and Small Fragment), IBI Scientific
Gel/PCR DNA Fragment Extraction Kits (Large and Small Fragment), IBI Scientific
Supplier:  IBI Scientific
Certificates
Gel/PCR DNA Fragment Extraction Kits (Large and Small Fragment), IBI Scientific
Supplier:  IBI Scientific
Specifications
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    DNA gel bands
    Amplified/modified DNA
  • Sample type:
    Gel matrix
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Small DNA Fragment Extraction Kit