HyClone™ Fetal Bovine Serum (United States), Super Low IgG, HyClone Products (Cytiva)
: Cytiva
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS).
- Processed via protein G chromatography
- Enhance cell culture growth
- U.S. sourced
Enhance cell culture growth with U.S.-sourced Cytiva HyClone Super Low IgG Fetal Bovine Serum (FBS), processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA76534-640
- CA76324-914
- CA95042-944
- CA95042-942
- CA76324-912
- CA95042-948
- Storage temperature:
- -
- -
- less than –10 °C
- less than –10 °C
- -
- less than –10 °C
Frequently Bought Together