Augment growth of cells with U.S.-sourced Cytiva HyClone Dialyzed Fetal Bovine Serum (FBS).
- Depleted of small molecules (less than 10000 MW)
Filtered through sequential 100 nm (0.1 μm) pore-size rated filters.
Dialysed foetal bovine serum (FBS) is used in applications requiring serum depleted of small molecules (<10000 MW) and is filtered through filtered through three sequential 100 nm (0,1 µm) pore-size related filters. The diafiltration process reduces concentrations of low molecular weight components, such as nucleotides and amino acids, that are necessary for alternative biochemical survival pathways. The process is reproducible and reduces hypoxanthine and thymidine concentrations below detectable limits. The glucose concentration, which is representative of the extent of dialysis, is reported for each lot of serum.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA76237-704
- CA16777-212
- CA76237-702
- CA76635-884
- CA16777-210
- CA76237-700
- Storage temperature:
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C
- Sera should be stored at –10 °C or lower. Once thawed, sera should be stored at 2...8 °C