Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Supplier: Scientific Plastics
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Supplier: Scientific Plastics
Certificates
Jump to:
About this item
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
- Beveled sides help reduce splash
- Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.
6 Options Available Below
Product Details & Documents
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
- Beveled sides help reduce splash
- Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.
Product Family Options
Product Information
- DésignationColorAvailabilityPrice
- Supplier Number: W39081-150
- Supplier Number: W39081-154
- Supplier Number: W39081-158
- Supplier Number: F164L081-150
- Supplier Number: F164L081-154
- Supplier Number: F164L081-158