Axygen® Gel Cutting Tips, Corning
Supplier: Corning
Certificates
About this item
Axygen® Gel Cutting Tips are designed to remove bands from agarose gel.
- Designed for removing bands from agarose gel
- One handed operation
- Push button gel release
- Ejector button tip release
- Avoid cross contamination
- Tips fit standard 1000 µL pipettors
These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
Frequently Bought Together