Axygen® Robotic Pipette Tips, ZT Series, Corning
89124-996
: Corning
Product available on GSA Advantage®
- Volume:30 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:89124-996
- Tip Style:384 Well Format
- Supplier No.:ZT-384-C-R
The ZT Series Robotic tips for the Zymark® RapidPlate/Cyclone automated systems are certified RNase-, DNase-, and pyrogen-free.
The ZT Series Robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.