Axygen® Gel Loading Tips, Corning
Axygen® Gel Loading Tips, Corning
Supplier:   Corning
Axygen® Gel Loading Tips, Corning
Supplier:  Corning
Certificates
Axygen® Gel Loading Tips, Corning
Axygen® Gel Loading Tips, Corning
4 Options Available Below
Product Details & Documents
Axygen brand gel loading tips have a universal fit for reseach grade pipettors.
They have a convenient graduation mark at 5 µl for quick volume check. The 0.17 mm tips are for loading 0.2 mm thick gels.
Product Family Options
Product Information
  • Tip Style
    Volume
    Sterile
    Tip Color
    Packed
    Availability
    Price
  • Flat, O.D. 0.017 cm (0.007")
    10 µl
    Clear
    200 Tips/Rack
    Log in for availability
    Log in for availability
    Case of 400
    Case of 400 - $2,560.00
    $2,560.00
    Case of 400
       $2,560.00
    Supplier Number: TGL-10FT-17R
  • Flat, O.D. 0.037 cm (0.015")
    200 µl
    Clear
    200 Tips/Rack
    Log in for availability
    Log in for availability
    Case of 400
    Case of 400 - $1,110.00
    $1,110.00
    Case of 400
       $1,110.00
    Supplier Number: TGL-200FT37R
  • Round, O.D. 0.057 cm (0.022")
    200 µl
    Clear
    200 Tips/Rack
    Log in for availability
    Log in for availability
    Case of 400
    Case of 400 - $140.25
    $140.25
    New Customer Promotional Price
    Case of 400
       $140.25
    Supplier Number: TGL-200RD-57-R
  • Round, O.D. 0.057 cm (0.022")
    200 µl
    +
    Clear
    200 Tips/Rack
    Log in for availability
    Log in for availability
    Case of 400
    Case of 400 - $156.00
    $156.00
    New Customer Promotional Price
    Case of 400
       $156.00
    Supplier Number: TGL-200RD-57-R-S