HyClone™ Bovine Growth Serum is a foetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
- Sourced in the USA with complete traceability back to original source
- Cost-effective alternative to FBS
- Growth promotion studies demonstrate that BGS is an excellent replacement for FBS
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
HyClone Bovine Growth Serum, U.S. Origin is supplemented with chemically defined components including amino acids, trace metals, vitamins, and other small molecules. Save money while ensuring excellent culture results with this cost-effective alternative to FBS. Bovine Growth Serum was developed for cell culturists who are concerned about the availability of fetal bovine serum and are seeking an affordable and viable alternative. BGS starts with Defined Bovine Calf Serum and is supplemented to stimulate cell growth and proliferation.
Origin is supplemented with chemically defined components including amino acids, trace metals, vitamins, and other small molecules.
Additional treatments available including heat-inactivation and irradiation. Additional testing, including EMEA-conforming testing, is also available.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA82006-906
- CA76324-910
- CA82013-586
- CA82006-908
- Origin:
- Animal
- Endotoxin:
- ≤10 EU/ml
- Hemoglobin:
- <30 mg/dl
- Storage temperature:
- Less than –10 °C
- −10 °C or lower. Once thawed, sera should be stored at 2..8 °C.
- Less than –10 °C
- Less than –10 °C