Automation Pipette Tips for PlateMate, PlateMate Plus, and Versette Workstations, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot.
Each lot must pass a 15-point QC inspection including performance testing on our PlateMate workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Frequently Bought Together