HyClone™ Phosphate Buffered Saline (PBS), HyClone products (Cytiva)
: Cytiva
Balanced salt solution used for multiple cell culture applications.
- Full traceability and documented origin of all formula ingredients
- Validated and consistent production processes
0.1 µm sterile filtered
Endotoxin Concentration: ≤1.0 EU/mL
: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Frequently Bought Together