Stainless Steel Closures, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These closures are designed for use with culture tubes and flasks and are available with or without gripping finger indents
Closures are made of 305 Stainless Steel and are fully autoclavable.
Frequently Bought Together