Flask, Fernbach, Baffled, Culture, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This Fernbach culture flask has a long-style 38mm neck for plastic or stainless steel closures
Triple baffles are located at the center of the bottom to achieve maximal oxygen transfer to culture medium. Fabricated from borosilicate glass.
Ordering information: Closure NOT included. Caution: Do not place on direct heat source.
Frequently Bought Together