Corning® Modified Impeller Assembly for Magnetically-Driven 3L Corning® Proculture® Spinner Flask, Corning
89090-178
: Corning
Product available on GSA Advantage®
- Description:Impeller assembly, stainless steel, dual bearing, for magnetically driven flask 3 L (to use with probes)
- Cat. no.:89090-178
- Fits Flask:3L angled sidearm spinner flasks (22877-080, 22877-090)
- Supplier No.:402648
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 3L Proculture® Spinner Flasks
- Narrower impeller paddle design allows use with probes to create a small bioreactor
- Non sterile
- Reusable