Zirconium ICP Standard, 1 ml = 1 mg Zr (1000 ppm Zr)ZrO(NO₃)₂ in 3% HNO₃, RICCA Chemical Company
Catalog # RCPZR1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. no.:RCPZR1KN-100
- Element:Zirconium
- Supplier No.:RPZR1KN0-100N