Gallium ICP Standard, 1 mL = 10 mg Ga (10,000 ppm Ga)Ga in 5% HNO3, Ricca Chemical Company
Catalogue # RCPGA10KN-50
Fournisseur: Ricca Chemical
Small Business Enterprise
Numéro CAS:
Spécifications
- Concentration:10000 µg/ml
- Matrix:5% HNO₃
- Pack type:Poly
- Size:50 ml
- Cat. no.:RCPGA10KN-50
- Element:Gallium
- Supplier No.:RPGA10KN-50N