Chromium(VI) AA Standard, 1 ml = 1 mg Cr (1,000 ppm Cr)(NH₄)₂Cr₂O₇ in 3% HNO₃, Ricca Chemical Company
RCACR61KN-500
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:500 ml
- Cat. no.:RCACR61KN-500
- Element:Chromium
- Supplier No.:RACR61KN-500N