Cryule® Vial Freezer Box | Avantor
Cryule® Vial Freezer Box
Cryule® Vial Freezer Box
Certificates
Cryule® Vial Freezer Box
Cryule® Vial Freezer Box
2 Options Available Below
Product Details & Documents
The WHEATON Cryule® Freezer box stores 1 and 2 mL cryogenic ampules and vials
Box dimensions are the following: 1 mL Cryule Freezer Box - 133 x 133 x 32 mm; 2 mL Cryule Freezer Box - 133 x 133 x 44 mm.
Made from polyethylene coated chipboard, it is designed for use in the vapor phase of liquid nitrogen. The box is designed to hold 81 freestanding or round bottom vials or ampules.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Dimensions
    Availability
    Price
  • Vial Freezer Box, 1 mL
    Red
    133×133×32 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 15
    Case of 15 - 302,60 $
    302,60 $
    Prix promotionnel pour les nouveaux clients
    Case of 15
       302,60 $
    Supplier Number: 651490
  • Vial Freezer Box, 2 mL
    Red
    133×133×44 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 15
    Case of 15 - 232,05 $
    232,05 $
    Prix promotionnel pour les nouveaux clients
    Case of 15
       232,05 $
    Supplier Number: 651492