Dropping Bottle, with Tip and Cap, LDPE, WHEATON®, DWK Life Sciences
WHEATON Dropping Bottles are made of LDPE (Low Density Polyethyleye)
The extended controlled dropper tip dispneses one drop at a time, with an average drop size of 44 µl ± 6 µl. Radiation grade polypropylene screw caps and tips packaged separately. Dropping bottles, natural tips and white caps are packaged separately.
Dropping bottles with tips can be used to dispense a stream of liquid or a non-controlled drop. Dropping bottles, tips and caps are packaged separately and bottles are autoclavable.
Frequently Bought Together