À propos de cet article
These Corning® slow-speed stirrers have one position with integral controls that can accommodate up to a ten or two-liter vessel.
Designed to be used with Corning® glass and plastic spinner flasks to provide optimum stirring speeds for both good mixing and high cell viability. Stirrers operate on 120V/60Hz and are UL approved to meet needs in North America and other locations with similar requirements. A compact design saves valuable bench space and offers powerful stirring from 10 to 150 rpm. Yet the economical features (just one stirring position) make it an ideal alternative to more expensive multiple position units. Rugged lightweight glass filled resin case has a low profile design. IP 65 rated. Can be used in a maximum 80% relative humidity environment. 2 year warranty.
Spécifications
- Catalog No:
- 89091-040
- W×D×H:
- 260×260×70 mm (1¼×10¼×2¾")