Zebra Fish Rearing System | Avantor
Zebra Fish Rearing System
Zebra Fish Rearing System, Benchtop Version, 50 1/4W x 54 1/2H x 14"D
Catalog # 470021-506
Supplier:   AQUANEERING LLC
Fournisseur #:  ZT350
Zebra Fish Rearing System
Catalog # 470021-506
Supplier:  AQUANEERING LLC
Zebra Fish Rearing System
Zebra Fish Rearing System
Zebra Fish Rearing System, Benchtop Version, 50 1/4W x 54 1/2H x 14"D
Price
$25,515.99
Log in pour la disponibilité
Each
Product Details & Documents
Perfect for Small-Scale Zebra Fish Educational Protocols.
  • Incorporates a biofilter design into a self-contained three-tier table top rack system
  • 250W heater maintains a comfortable environment throughout the rack
  • Tanks are self-cleaning
Spend less time maintaining your zebra fish and more time on the actual lab with this low-maintenance housing system. It incorporates a biofilter design into a self-contained three-tier table top rack system. The filtration system performs mechanical, chemical, and biological filtration, as well as UV sterilization so the water is recirculated and clean with undetectable levels of ammonia. The 250 W heater maintains a comfortable environment throughout the rack, which can house sixteen 1.0 L, twelve 2.8 L, and five 9.0 L polycarbonate tanks in varying configurations. All of the tanks mentioned above are self-cleaning and are included in the system. The rack is stainless steel with adjustable legs.
Delivery information: This item ships freight and may have additional shipping charges or require a loading dock.
Spécifications
  • Désignation:
    Zebra Fish Rearing System, Benchtop Version
  • Dimensions:
    50 1/4W x 54 1/2H x 14"D