Zebra Fish Development Slide Set
Zebra Fish Development Slide Set, 6, Cellular Development
Catalog # 470183-526
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Vertebrate Development
- Identifying features clearly distinguishable
- Rigorous quality control standards
Prepared in Ward's® own labs by skilled technicians, the slides show the detailed anatomy of organisms commonly used in genetic and developmental studies.
Ordering information: Set includes Cleavage stage (wm); Embryo stage (wm); Hatchling stage (wm); Adult female (ls); Adult male (ls); and Adult, composite (cs).
Specifications
- Type:Cellular Development
- Désignation:Zebra Fish Development Slide Set
- No. of Slides:6