Z Stool
Supplier: National Public Seating
À propos de cet article
Lengthy labs require students to be alert, focused, and free of distraction from uncomfortable seating. These stylish stools keep students comfortable during long labs.
- Ergonomic blow molded contoured for increased air circulation
- Attractive, 12-gauge, chrome-plated steel frame and rear brace add strength and durability
- U-shaped base and Z-shaped legs
- Stools sweep across all floor types with permanently attached glides
- Nests on tabletops
Stools are available in three heights, with or without backs, and are easily stackable for convenient storage.
11 Options Available Below
Product Details & Documents
Lengthy labs require students to be alert, focused, and free of distraction from uncomfortable seating. These stylish stools keep students comfortable during long labs.
- Ergonomic blow molded contoured for increased air circulation
- Attractive, 12-gauge, chrome-plated steel frame and rear brace add strength and durability
- U-shaped base and Z-shaped legs
- Stools sweep across all floor types with permanently attached glides
- Nests on tabletops
Stools are available in three heights, with or without backs, and are easily stackable for convenient storage.
Specifications for Product Family
Spécifications
- Catalog No:
- 470128-910
- 76276-942
- 470128-912
- 76276-944
- 470128-914
- 76276-634
- 470128-916
- 470128-918
- 76276-938
- 470128-920
- 76276-940
- Model:
- 470128-910
- 76276-942
- 470128-912
- 76276-944
- 470128-914
- 76276-634
- 470128-916
- 470128-918
- 76276-938
- 470128-920
- 76276-940
- Frame colour/finish:
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Frame material:
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Seat/back color:
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Seat/back material:
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Plastic
- Backrest dimensions:
- W×H 24"
- W×H 24"
- W×H 24"
- W×H 24"
- W×H 24"
- W×H 24"
Spécifications
Product Family Options
Informations sur le produit
- DésignationColorFoot StyleSeat HeightAvailabilityPrice
- Supplier Number: 6618BSpécifications:
More
Specifications
Cat. No.470128-910Model470128-910Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensionsW×H 24" - Supplier Number: 6618BSpécifications:
More
Specifications
Cat. No.76276-942Model76276-942Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensionsW×H 24" - Supplier Number: 6622BSpécifications:
More
Specifications
Cat. No.470128-912Model470128-912Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensionsW×H 24" - Supplier Number: 6622BSpécifications:
More
Specifications
Cat. No.76276-944Model76276-944Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensionsW×H 24" - Supplier Number: 6624BSpécifications:
More
Specifications
Cat. No.470128-914Model470128-914Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensionsW×H 24" - Supplier Number: 6624BSpécifications:
More
Specifications
Cat. No.76276-634Model76276-634Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensionsW×H 24" - Supplier Number: 6618Spécifications:
More
Specifications
Cat. No.470128-916Model470128-916Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensions - Supplier Number: 6622Spécifications:
More
Specifications
Cat. No.470128-918Model470128-918Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensions - Supplier Number: 6622Spécifications:
More
Specifications
Cat. No.76276-938Model76276-938Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensions - Supplier Number: 6624Spécifications:
More
Specifications
Cat. No.470128-920Model470128-920Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensions - Supplier Number: 6624Spécifications:
More
Specifications
Cat. No.76276-940Model76276-940Frame colour/finishChromeFrame materialSteelSeat/back colorBlackSeat/back materialPlasticBackrest dimensions