Wraparound Adjustable Safety Glasses
Catalogue # 470017-066
Fournisseur: VWR International
Spécifications
- Description:Wraparound safety glasses, adjustable
- Cat. no.:470017-066
- Supplier No.:ET-83R-2
Spécifications
À propos de cet article
With these stylish glasses, safety doesn't have to be boring.
- Impact-resistant polycarbonate lenses
- Glasses provide 99.9% UV protection
- Unisex
All models feature an ultra-light wraparound design and are suitable for men and women.
Certifications: Meet ANSI Z87+ Specifications.