Whose DNA Was Left Behind?
Catalog # 470200-480
Supplier: Edvotek
Specifications
- Description:Whose DNA Was Left Behind Kit
- No. of Lab Groups:10
- Cat. no.:470200-480
- Supplier No.:S-51
Specifications
About this item
Students identify individuals based on DNA analysis.
- Convenient, Ready-To-Load Samples
- Time required: 45 minutes
- Students Identify Individuals Based on DNA Analysis
- Electrophoresis Buffer Included
DNA obtained from a single hair left behind at a crime scene can be used to identify a criminal.
In this 45 minute experiment, students compare simulated crime scene DNA with that of two suspects.
Ordering information: This kit includes Ready-to-Load dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes. Electrophoresis apparatus, a power supply, and a microwave or hot plate are required but not included.
Delivery: Store Ready-to-Load QuickStrip samples in the refrigerator after they are delivered.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator.