White Plastic Meter Stick
Metric meter stick, White, Plastic, 1 m
Catalog # 470047-180
Supplier: SI Manufacturing
Fournisseur #: 11000-B
Jump to:
Product Details & Documents
Thick Plastic Ideal for Classroom Use.
- Solid 6 mm thick plastic
- Calibrated on one side in mm
- Other side calibrated in 0.5 cm increments
- Aligned with GLOBE protocols
Spécifications
- Color:White
- Désignation:Metric meter stick
- Longueur:1 m
- Matière:Plastic